Assay Details
Gene Symbol: DNAJB4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001498098.4 | XP_001498148.1 | 1169 | 157 | ||
XM_005610480.3 | XP_005610537.1 | 1148 | 157 | |||
XM_023641816.1 | XP_023497584.1 | 893 | 157 | |||
XM_023641817.1 | XP_023497585.1 | 906 | 157 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
