Assay Details
Gene Symbol: SERBP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023641883.1 | XP_023497651.1 | 1303 | 59 | ||
XM_023641884.1 | XP_023497652.1 | 1285 | 59 | |||
XM_023641885.1 | XP_023497653.1 | 1258 | 59 | |||
XM_023641886.1 | XP_023497654.1 | 1240 | 59 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
