Assay Details
Gene Symbol: AK4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005610525.3 | XP_005610582.1 | 1022 | 99 | ||
XM_005610526.2 | XP_005610583.1 | 733 | 99 | |||
XM_023641944.1 | XP_023497712.1 | 915 | 99 | |||
XM_023641945.1 | XP_023497713.1 | 795 | 99 | |||
XM_023641947.1 | XP_023497715.1 | 781 | 99 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
