Assay Details
Gene Symbol: NUDT22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001489010.5 | XP_001489060.4 | 865 | 64 | ||
XM_023654284.1 | XP_023510052.1 | 877 | 64 | |||
XM_023654285.1 | XP_023510053.1 | 812 | 64 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
