Assay Details
Gene Symbol: BAD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001916754.3 | XP_001916789.1 | 343 | 98 | ||
XM_023654304.1 | XP_023510072.1 | 220 | 98 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: BAD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001916754.3 | XP_001916789.1 | 343 | 98 | ||
XM_023654304.1 | XP_023510072.1 | 220 | 98 |