Assay Details
Gene Symbol: BAD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001916754.3 | XP_001916789.1 | 532 | 61 | ||
XM_023654304.1 | XP_023510072.1 | 409 | 61 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
