Assay Details
Gene Symbol: GRK2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023654544.1 | XP_023510312.1 | 1032 | 63 | ||
XM_023654545.1 | XP_023510313.1 | 1033 | 63 |

More Information
Set Membership: |
![]() ![]() ![]() |
