Assay Details
Gene Symbol: POLD4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001497352.4 | XP_001497402.3 | 289 | 79 | ||
XM_023654559.1 | XP_023510327.1 | 289 | 79 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
