Assay Details
Gene Symbol: NDUFS8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001498426.5 | XP_001498476.3 | 490 | 62 | ||
XM_023654586.1 | XP_023510354.1 | 397 | 62 |

More Information
Set Membership: |
![]() ![]() ![]() |
