Assay Details
Gene Symbol: DHCR7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023654657.1 | XP_023510425.1 | 1061 | 65 | ||
XM_023654658.1 | XP_023510426.1 | 995 | 65 |

More Information
Set Membership: |
![]() ![]() ![]() |
