Assay Details
Gene Symbol: MRPL23 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023654703.1 | XP_023510471.1 | 826 | 60 | ||
XM_023654704.1 | XP_023510472.1 | 743 | 60 | |||
XM_023654705.1 | XP_023510473.1 | 433 | 60 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
