Assay Details
Gene Symbol: HRAS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023654737.1 | XP_023510505.1 | 296 | 81 | ||
XM_023654738.1 | XP_023510506.1 | 296 | 81 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: HRAS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023654737.1 | XP_023510505.1 | 296 | 81 | ||
XM_023654738.1 | XP_023510506.1 | 296 | 81 |