Assay Details
Gene Symbol: DUSP8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023654827.1 | XP_023510595.1 | 6569 | 79 | ||
XM_023654828.1 | XP_023510596.1 | 876 | 79 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: DUSP8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023654827.1 | XP_023510595.1 | 6569 | 79 | ||
XM_023654828.1 | XP_023510596.1 | 876 | 79 |