Assay Details
Gene Symbol: PTCD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005598516.3 | XP_005598573.1 | 956 | 76 | ||
XM_014729798.2 | XP_014585284.1 | 959 | 76 | |||
XM_023655298.1 | XP_023511066.1 | 961 | 76 | |||
XM_023655299.1 | XP_023511067.1 | 849 | 76 | |||
XM_023655300.1 | XP_023511068.1 | 747 | 76 |

More Information
Set Membership: |
![]() ![]() ![]() |
