Assay Details
Gene Symbol: RHBDD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023655423.1 | XP_023511191.1 | 673 | 76 | ||
XM_023655424.1 | XP_023511192.1 | 673 | 76 |

More Information
Set Membership: |
![]() ![]() ![]() |
