Assay Details
Gene Symbol: RHBDD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023655424.1 | XP_023511192.1 | 833 | 69 | ||
XM_023655425.1 | XP_023511193.1 | 402 | 69 | |||
XM_023655426.1 | XP_023511194.1 | 366 | 69 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
