Assay Details
Gene Symbol: BUD23 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001504510.4 | XP_001504560.1 | 605 | 56 | ||
XM_023655451.1 | XP_023511219.1 | 725 | 56 | |||
XM_023655453.1 | XP_023511221.1 | 725 | 56 | |||
XM_023655454.1 | XP_023511222.1 | 725 | 56 |

More Information
Set Membership: |
![]() ![]() ![]() |
