Assay Details
Gene Symbol: FUS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005598742.3 | XP_005598799.1 | 461 | 65 | ||
XM_005598743.3 | XP_005598800.1 | 458 | 65 |

More Information
Set Membership: |
![]() ![]() ![]() |
