Assay Details
Gene Symbol: MAZ | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023616000.1 | XP_023471768.1 | 1192 | 66 | ||
XM_023616001.1 | XP_023471769.1 | 1163 | 66 | |||
XM_023616002.1 | XP_023471770.1 | 1191 | 66 | |||
XM_023616003.1 | XP_023471771.1 | 1148 | 66 | |||
XM_023616004.1 | XP_023471772.1 | 1118 | 66 | |||
XM_023616005.1 | XP_023471773.1 | 1148 | 66 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
