Assay Details
Gene Symbol: KCTD13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_014730024.2 | XP_014585510.2 | 725 | 108 | ||
XM_023616011.1 | XP_023471779.1 | 638 | 108 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
