Assay Details
Gene Symbol: ALDOA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003362712.3 | XP_003362760.1 | 587 | 54 | ||
XM_003362713.4 | XP_003362761.1 | 538 | 54 | |||
XM_005598800.3 | XP_005598857.1 | 520 | 54 | |||
XM_005598801.3 | XP_005598858.1 | 526 | 54 | |||
XM_005598802.2 | XP_005598859.1 | 422 | 54 | |||
XM_023616037.1 | XP_023471805.1 | 554 | 54 |

More Information
Set Membership: |
![]() ![]() ![]() |
