Assay Details
Gene Symbol: NDUFAB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023616118.1 | XP_023471886.1 | 462 | 109 | ||
XM_023616119.1 | XP_023471887.1 | 329 | 109 |

More Information
Set Membership: |
![]() ![]() |
