Assay Details
Gene Symbol: NDUFAB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023616118.1 | XP_023471886.1 | 548 | 89 | ||
XM_023616119.1 | XP_023471887.1 | 415 | 89 |

More Information
Set Membership: |
![]() ![]() ![]() |
