Assay Details
Gene Symbol: TVP23A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005598922.3 | XP_005598979.1 | 1260 | 65 | ||
XM_023616323.1 | XP_023472091.1 | 1351 | 65 | |||
XM_023616324.1 | XP_023472092.1 | 1352 | 65 | |||
XM_023616325.1 | XP_023472093.1 | 1352 | 65 | |||
XM_023616326.1 | XP_023472094.1 | 1353 | 65 |

More Information
Set Membership: |
![]() ![]() ![]() |
