Assay Details
Gene Symbol: CDIP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005598995.3 | XP_005599052.1 | 475 | 89 | ||
XM_005598996.3 | XP_005599053.1 | 465 | 89 | |||
XM_005598997.3 | XP_005599054.1 | 914 | 89 | |||
XM_005598998.3 | XP_005599055.1 | 791 | 89 | |||
XM_005599001.3 | XP_005599058.1 | 477 | 89 | |||
XM_014730187.2 | XP_014585673.1 | 664 | 89 | |||
XM_023616405.1 | XP_023472173.1 | 768 | 89 | |||
XM_023616406.1 | XP_023472174.1 | 403 | 89 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
