Assay Details
Gene Symbol: HMOX2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001502330.6 | XP_001502380.1 | 976 | 73 | ||
XM_014730185.2 | XP_014585671.1 | 1004 | 73 | |||
XM_023616404.1 | XP_023472172.1 | 1006 | 73 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
