Assay Details
Gene Symbol: PKMYT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001499512.4 | XP_001499562.1 | 1418 | 76 | ||
XM_005599045.3 | XP_005599102.1 | 1507 | 76 | |||
XM_005599046.3 | XP_005599103.1 | 1508 | 76 | |||
XM_023616503.1 | XP_023472271.1 | 1310 | 76 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
