Assay Details
Gene Symbol: CRAMP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_014730258.2 | XP_014585744.2 | 2434 | 66 | ||
XM_023616636.1 | XP_023472404.1 | 1018 | 66 | |||
XM_023616638.1 | XP_023472406.1 | 1237 | 66 |

More Information
Set Membership: |
![]() ![]() ![]() |
