Assay Details
Gene Symbol: CCDC154 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005599072.3 | XP_005599129.1 | 1818 | 71 | ||
XM_023616658.1 | XP_023472426.1 | 2033 | 71 | |||
XM_023616659.1 | XP_023472427.1 | 1818 | 71 |
