Assay Details
Gene Symbol: MPG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001494795.6 | XP_001494845.3 | 180 | 109 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_001494795.6 transcripts.
Set Membership: |
![]() ![]() |
