Assay Details
Gene Symbol: HNRNPAB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023617077.1 | XP_023472845.1 | 910 | 97 | ||
XM_023617078.1 | XP_023472846.1 | 910 | 97 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
