Assay Details
Gene Symbol: MXD3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001502408.4 | XP_001502458.2 | 717 | 58 | ||
XM_023617137.1 | XP_023472905.1 | 425 | 58 |

More Information
Set Membership: |
![]() ![]() ![]() |
