Assay Details
Gene Symbol: EIF4E1B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005599205.3 | XP_005599262.1 | 423 | 60 | ||
XM_023617168.1 | XP_023472936.1 | 424 | 60 |

More Information
Set Membership: |
![]() ![]() ![]() |
