Assay Details
Gene Symbol: CPEB4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001502804.6 | XP_001502854.1 | 2695 | 63 | ||
XM_005599224.3 | XP_005599281.1 | 2670 | 63 | |||
XM_005599225.3 | XP_005599282.1 | 2619 | 63 | |||
XM_023617218.1 | XP_023472986.1 | 2644 | 63 |

More Information
Set Membership: |
![]() ![]() ![]() |
