Assay Details
Gene Symbol: BNIP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023617222.1 | XP_023472990.1 | 556 | 82 | ||
XM_023617223.1 | XP_023472991.1 | 679 | 82 |

More Information
Set Membership: |
![]() ![]() ![]() |
