Assay Details
Gene Symbol: PANK3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023617272.1 | XP_023473040.1 | 1121 | 67 | ||
GenBank mRNA | JL630672.1 | - | 3 - 4 | 503 | 67 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
