Assay Details
Gene Symbol: GABRB2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001503408.4 | XP_001503458.1 | 601 | 73 | ||
XM_005599237.3 | XP_005599294.1 | 595 | 73 | |||
XM_023617283.1 | XP_023473051.1 | 521 | 73 | |||
XM_023617285.1 | XP_023473053.1 | 497 | 73 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
