Assay Details
Gene Symbol: CCNJL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005599251.3 | XP_005599308.2 | 1061 | 64 | ||
XM_023617300.1 | XP_023473068.1 | 1093 | 64 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
