Assay Details
Gene Symbol: GRPEL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001503782.4 | XP_001503832.1 | 436 | 104 | ||
XM_023617461.1 | XP_023473229.1 | 490 | 104 | |||
GenBank mRNA | JL616140.1 | - | 2 - 3 | 187 | 104 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
