Assay Details
Gene Symbol: SPINK1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003362792.4 | XP_003362840.1 | 200 | 136 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_003362792.4 transcripts.
Set Membership: |
![]() ![]() |
