Assay Details
Gene Symbol: GFRA3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001502575.5 | XP_001502625.2 | 1393 | 70 | ||
XM_014730647.2 | XP_014586133.1 | 1393 | 70 | |||
XM_023617629.1 | XP_023473397.1 | 1361 | 70 | |||
XM_023617630.1 | XP_023473398.1 | 1300 | 70 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
