Assay Details
Gene Symbol: VDAC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005599430.3 | XP_005599487.1 | 669 | 76 | ||
XM_023617695.1 | XP_023473463.1 | 671 | 76 |

More Information
Set Membership: |
![]() ![]() ![]() |
