Assay Details
Gene Symbol: RPL7A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023629241.1 | XP_023485009.1 | 371 | 77 | ||
GenBank mRNA | AF508309.1 | - | 2 - 3 | 170 | 77 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
