Assay Details
Gene Symbol: KCNJ9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005615223.2 | XP_005615280.2 | 4318 | 78 | ||
XM_023640903.1 | XP_023496671.1 | 2057 | 78 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
