Assay Details
Gene Symbol: AUP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001916596.4 | XP_001916631.3 | 874 | 54 | ||
XM_014731078.2 | XP_014586564.1 | 879 | 54 | |||
XM_014731079.2 | XP_014586565.1 | 878 | 54 |

More Information
Set Membership: |
![]() ![]() ![]() |
