Assay Details
Gene Symbol: RELL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005599341.3 | XP_005599398.1 | 1434 | 69 | ||
XM_023617525.1 | XP_023473293.1 | 1441 | 69 | |||
XM_023617526.1 | XP_023473294.1 | 1190 | 69 | |||
XM_023617527.1 | XP_023473295.1 | 1197 | 69 |

More Information
Set Membership: |
![]() ![]() ![]() |
