Assay Details
Gene Symbol: NOXRED1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005605379.3 | XP_005605436.1 | 1130 | 73 | ||
XM_005605380.3 | XP_005605437.1 | 818 | 73 | |||
XM_023628176.1 | XP_023483944.1 | 1161 | 73 | |||
XM_023628177.1 | XP_023483945.1 | 1126 | 73 | |||
XM_023628178.1 | XP_023483946.1 | 801 | 73 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
