Assay Details
Gene Symbol: LOC100060483 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005607698.3 | XP_005607755.1 | 169 | 79 | ||
XM_005607699.3 | XP_005607756.1 | 169 | 79 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_005607698.3, XM_005607699.3 transcripts.
Set Membership: |
![]() ![]() |
