Assay Details
Gene Symbol: DNASE1L2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023616556.1 | XP_023472324.1 | 823 | 59 | ||
XM_023616557.1 | XP_023472325.1 | 820 | 59 |

More Information
Set Membership: |
![]() ![]() ![]() |
