Assay Details
Gene Symbol: NDUFS3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001491272.6 | XP_001491322.2 | 580 | 61 | ||
XM_023654014.1 | XP_023509782.1 | 580 | 61 |

More Information
Set Membership: |
![]() ![]() ![]() |
